Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psma3
Entrez Gene ID
19167
Gene Name
proteasome (prosome, macropain) subunit, alpha type 3
Gene Aliases
Lmpc8
Chromosome Location
Chr.12: 70974623 - 70994937 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011184.4 NP_035314.3 2 - 3 223 77
GenBank mRNA AF055983.1 - 2 - 3 141 77
AF060089.1 - 2 - 3 109 77
AI303320.1 - 2 - 3 222 77
AK002877.1 - 2 - 4 77
AK165424.1 - 2 - 3 176 77
AK168495.1 - 2 - 3 128 77
BC091743.1 - 2 - 3 113 77

View Full Product Details