Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gnpnat1
Entrez Gene ID
54342
Gene Name
glucosamine-phosphate N-acetyltransferase 1
Gene Aliases
AU017428, AU040593, EMeg32, Gpnat1, Gsnpat
Chromosome Location
Chr.14: 45376421 - 45388956 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019425.2 NP_062298.1 5 - 6 470 102
GenBank mRNA AJ001006.1 - 5 - 6 470 102
AK002466.1 - 5 - 6 491 102
AK007647.1 - 5 - 6 475 102
AK007722.1 - 5 - 6 475 102
AK007764.1 - 5 - 6 498 102
AK008566.1 - 5 - 6 480 102
AK011098.1 - 5 - 6 498 102
AK018499.1 - 5 - 6 464 102
AK050266.1 - 5 - 6 468 102
AK167870.1 - 5 - 6 480 102
AK168074.1 - 5 - 6 480 102
BC031116.1 - 5 - 6 460 102
BC092047.1 - 5 - 6 451 102

View Full Product Details