The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023434.3 | NP_075923.2 | 6 - 7 | 1160 | 187 |
GenBank mRNA | AF228408.1 | - | 4 - 5 | 819 | 187 |
AK041829.1 | - | 3 - 4 | 2034 | 187 | |
AK042459.1 | - | 3 - 4 | 2034 | 187 | |
AK088144.1 | - | 5 - 6 | 832 | 187 | |
AK147023.1 | - | 5 - 6 | 866 | 187 | |
AK147834.1 | - | 5 - 6 | 837 | 187 | |
AK153278.1 | - | 5 - 6 | 826 | 187 | |
AK173012.1 | - | 5 - 6 | 864 | 187 | |
BC050091.1 | - | 4 - 5 | 815 | 187 |