The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026021.3 | NP_080297.1 | 4 - 5 | 552 | 165 |
GenBank mRNA | AJ409150.1 | - | 4 - 5 | 513 | 165 |
AJ409151.1 | - | 4 - 5 | 398 | 165 | |
AK005908.1 | - | 4 - 5 | 540 | 165 | |
AK012484.1 | - | 4 - 5 | 553 | 165 | |
AK088063.1 | - | 4 - 5 | 573 | 164 | |
BC027321.1 | - | 4 - 5 | 518 | 165 |