The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110129.1 | NP_001103599.1 | 2 - 3 | 181 | 74 |
NM_001110130.1 | NP_001103600.1 | 2 - 3 | 181 | 74 | |
NM_028677.4 | NP_082953.1 | 2 - 3 | 181 | 74 | |
GenBank mRNA | AK003179.1 | - | 2 - 3 | 159 | 74 |
AK005202.1 | - | 2 - 3 | 142 | 74 | |
AK008394.1 | - | 2 - 3 | 159 | 74 | |
AK014665.1 | - | 2 - 3 | 142 | 74 | |
AK134995.1 | - | 2 - 3 | 161 | 74 | |
AK136330.1 | - | 2 - 3 | 140 | 74 | |
AK164299.1 | - | 2 - 3 | 146 | 74 | |
BC016565.1 | - | 2 - 3 | 74 | ||
BC050116.1 | - | 2 - 3 | 135 | 74 | |
BC098490.1 | - | - | 74 | ||
BY708305.1 | - | 2 - 3 | 158 | 74 | |
CJ071503.1 | - | 2 - 3 | 204 | 74 |