Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aff1
Entrez Gene ID
17355
Gene Name
AF4/FMR2 family, member 1
Gene Aliases
9630032B01Rik, AW319193, Af4, Mllt2h, Rob
Chromosome Location
Chr.5: 103692334 - 103855322 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080798.1 NP_001074267.1 17 - 18 3325 150
NM_133919.3 NP_598680.3 17 - 18 3336 150
GenBank mRNA AF013131.1 - 16 - 17 3151 150
AF074266.1 - 17 - 18 3166 150
AY749168.1 - 17 - 18 3169 150
BC131665.1 - 17 - 18 3169 150
CN537099.1 - 4 - 5 334 150

View Full Product Details