Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmn1r5
Entrez Gene ID
171192
Gene Name
vomeronasal 1 receptor 5
Gene Aliases
V1rc19
Chromosome Location
Chr.6: 56985342 - 56986289 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134174.2 NP_598935.2 1 - 1 159 213
GenBank mRNA BC138068.1 - 1 - 1 206 213
BC138069.1 - 1 - 1 206 213

View Full Product Details