Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
D10Jhu81e
Entrez Gene ID
28295
Gene Name
DNA segment, Chr 10, Johns Hopkins University 81 expressed
Gene Aliases
C21orf33, ES1
Chromosome Location
Chr.10: 78162067 - 78169768 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138601.2 NP_613067.1 2 - 3 230 88
GenBank mRNA AK003866.1 - 2 - 3 231 88
BC013475.1 - 2 - 3 223 88

View Full Product Details