Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ints5
Entrez Gene ID
109077
Gene Name
integrator complex subunit 5
Gene Aliases
1110055N21Rik, mKIAA1698
Chromosome Location
Chr.19: 8892987 - 8897890 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176843.3 NP_789813.1 1 - 2 162 117
GenBank mRNA AK143868.1 - 1 - 2 124 117
AK144770.1 - 1 - 2 137 117
AK155499.1 - 1 - 2 124 117
AK166149.1 - 1 - 2 135 117
AK168795.1 - 1 - 2 135 117
AK170584.1 - 1 - 2 124 117
AK172260.1 - 1 - 2 124 117
BC039214.1 - 1 - 2 144 117
BY152900.1 - 1 - 2 117

View Full Product Details