The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178017.1 | NP_821136.1 | 10 - 11 | 1860 | 196 |
GenBank mRNA | AK088758.1 | - | 6 - 7 | 686 | 196 |
BC023167.1 | - | 6 - 7 | 602 | 196 | |
BC048188.1 | - | 11 - 12 | 1975 | 196 | |
BC049800.1 | - | 10 - 11 | 1860 | 196 |