Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Esrp1
- Entrez Gene ID
-
207920
- Gene Name
- epithelial splicing regulatory protein 1
- Gene Aliases
-
2210008M09Rik, A630065D16, BC031468, Rbm35a
- Chromosome Location
-
Chr.4: 11331933 - 11386783 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC031468.1 |
- |
10 - 11 |
1288 |
74 |
View Full Product Details