The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | AK051219.1 | - | 7 - 8 | 1249 | 71 |
AK079571.1 | - | 7 - 8 | 1231 | 71 | |
AK079639.1 | - | 7 - 8 | 1230 | 71 | |
AK086262.1 | - | 7 - 8 | 1231 | 71 | |
AK140932.1 | - | 7 - 8 | 1269 | 71 | |
AK148462.1 | - | 7 - 8 | 1231 | 71 | |
AK159246.1 | - | 7 - 8 | 1289 | 71 | |
AK163040.1 | - | 7 - 8 | 1230 | 71 | |
AK169157.1 | - | 7 - 8 | 1230 | 71 | |
AK170195.1 | - | 7 - 8 | 1250 | 71 | |
BC003797.1 | - | 7 - 8 | 1191 | 71 | |
U14390.1 | - | 7 - 8 | 1221 | 71 |