Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dynlt1b
Entrez Gene ID
21648
Gene Name
dynein light chain Tctex-type 1B
Gene Aliases
AGS2, Dynlt1, Tctex-1, Tctex1
Chromosome Location
Chr.17: 6430112 - 6436296 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009342.2 NP_033368.1 2 - 3 167 75
GenBank mRNA AK133788.1 - 2 - 3 97 75
AK153971.1 - 2 - 3 102 75
BC043018.1 - 2 - 3 167 75
BC089472.1 - 2 - 3 111 75
M25825.1 - - 79 75

Gene Symbol
Dynlt1a
Entrez Gene ID
100310872
Gene Name
dynein light chain Tctex-type 1A
Gene Aliases
-
Chromosome Location
Chr.17: 6306344 - 6317474 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166629.2 NP_001160101.1 2 - 3 141 75
GenBank mRNA AK030569.1 - 2 - 3 102 75
BC087868.1 - 2 - 3 141 75

Gene Symbol
Dynlt1c
Entrez Gene ID
100040563
Gene Name
dynein light chain Tctex-type 1C
Gene Aliases
100040603, Dynlt1d
Chromosome Location
Chr.17: 6601779 - 6607891 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166630.1 NP_001160102.1 2 - 3 98 75
GenBank mRNA AK168478.1 - 2 - 3 99 75
EF110999.1 - - 70 75

Gene Symbol
Dynlt1f
Entrez Gene ID
100040531
Gene Name
dynein light chain Tctex-type 1F
Gene Aliases
100040631, Dynlt1e
Chromosome Location
Chr.17: 6646606 - 6655939 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166627.1 NP_001160099.1 2 - 3 206 75
NM_001199948.1 NP_001186877.1 2 - 3 206 75
GenBank mRNA AK005569.1 - 2 - 3 86 75
AK014409.1 - 2 - 3 86 75
BY099269.1 - 2 - 3 99 75
BY250333.1 - 2 - 3 206 75

View Full Product Details