Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Klrc2
- Entrez Gene ID
-
16642
- Gene Name
- killer cell lectin-like receptor subfamily C, member 2
- Gene Aliases
-
NKG2C
- Chromosome Location
-
Chr.6: 129649602 - 129660737 on Build GRCm38
- Gene Symbol
- Klrc3
- Entrez Gene ID
-
58179
- Gene Name
- killer cell lectin-like receptor subfamily C, member 3
- Gene Aliases
-
Klrc2, Nkg2e
- Chromosome Location
-
Chr.6: 129636263 - 129643511 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC130234.1 |
- |
2 - 3 |
269 |
116 |
View Full Product Details