Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spry1
Entrez Gene ID
24063
Gene Name
sprouty homolog 1 (Drosophila)
Gene Aliases
sprouty1, spry-1
Chromosome Location
Chr.3: 37639846 - 37644599 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF176903.1 - 1 - 2 139 82
AK014578.1 - 1 - 2 152 82
AK140405.1 - 1 - 2 151 82
BC039139.1 - 1 - 2 157 82

View Full Product Details