The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016687.3 | NP_057896.1 | 4 - 5 | 1050 | 115 |
GenBank mRNA | AF117709.1 | - | 4 - 5 | 1065 | 115 |
AK009981.1 | - | 4 - 5 | 1041 | 115 | |
AK080766.1 | - | 4 - 5 | 1051 | 115 |