The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277273.1 | NP_001264202.1 | 5 - 6 | 509 | 70 |
NM_025923.3 | NP_080199.1 | 5 - 6 | 509 | 70 | |
NR_102382.1 | 5 - 6 | 509 | 70 | ||
GenBank mRNA | AA476004.1 | - | 3 - 4 | 180 | 70 |
AF513619.1 | - | 5 - 6 | 509 | 70 | |
AK008605.1 | - | 5 - 6 | 395 | 70 | |
AK011520.1 | - | 5 - 6 | 414 | 70 | |
AK011752.1 | - | 5 - 6 | 386 | 70 | |
AK011903.1 | - | 5 - 6 | 392 | 70 | |
AK018018.1 | - | 5 - 6 | 391 | 70 | |
AK076017.1 | - | 5 - 6 | 392 | 70 | |
AK168538.1 | - | 5 - 6 | 393 | 70 | |
BC032876.1 | - | 5 - 6 | 383 | 70 | |
BC066181.1 | - | 5 - 6 | 438 | 70 | |
CN455644.1 | - | 4 - 5 | 295 | 70 |