The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138661.1 | NP_619602.1 | 1 - 2 | 2479 | 85 |
GenBank mRNA | AB008183.1 | - | 1 - 2 | 2282 | 85 |
AY013767.1 | - | 1 - 2 | 2479 | 85 |