Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Jmjd4
Entrez Gene ID
194952
Gene Name
jumonji domain containing 4
Gene Aliases
6430559I23
Chromosome Location
Chr.11: 59450045 - 59458567 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205068.1 NP_001191997.1 2 - 3 634 76
NM_178659.6 NP_848774.1 2 - 3 634 76
NR_037992.1 2 - 3 634 76
GenBank mRNA AK032477.1 - 2 - 3 608 76
AK037954.1 - 2 - 3 633 76
AK038650.1 - 1 - 2 177 76
AK039116.1 - 2 - 3 312 76
AK047586.1 - 2 - 3 635 76
AK052144.1 - 2 - 3 620 76
AK079104.1 - 2 - 3 592 76
AK084669.1 - 2 - 3 592 76
AK145394.1 - 2 - 3 612 76
AK171919.1 - 2 - 3 589 76
BC049148.1 - 2 - 3 619 76
BC116707.1 - 2 - 3 565 76
BC118020.1 - 2 - 3 564 76

View Full Product Details