Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Vmn2r37
- Entrez Gene ID
-
22305
- Gene Name
- vomeronasal 2, receptor 37
- Gene Aliases
-
V2r14
- Chromosome Location
-
Chr.7: 9205549 - 9223653 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_009489.2 |
NP_033515.2 |
2 - 3 |
598 |
153 |
GenBank mRNA |
AF011424.1 |
- |
2 - 3 |
598 |
153 |
- Gene Symbol
- Vmn2r34
- Entrez Gene ID
-
100042636
- Gene Name
- vomeronasal 2, receptor 34
- Gene Aliases
-
ENSMUSG00000070841
- Chromosome Location
-
Chr.7: 7671829 - 7689398 on Build GRCm38
View Full Product Details