The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020596.2 | NP_065621.1 | 1 - 2 | 282 | 172 |
GenBank mRNA | AK141637.1 | - | 1 - 2 | 329 | 172 |