Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gbp6
Entrez Gene ID
100702
Gene Name
guanylate binding protein 6
Gene Aliases
AI595338, Mpa2l
Chromosome Location
Chr.5: 105270702 - 105295870 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194336.2 NP_919317.2 10 - 11 1733 113
GenBank mRNA AK128993.1 - - 1729 113
AK129021.1 - - 1005 113
AK171913.1 - 7 - 8 3371 113
BC007143.1 - 9 - 10 1590 113
BC052867.1 - 5 - 6 961 113
BC057969.1 - - 1719 113
BC115768.1 - - 1729 113

View Full Product Details