Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krtap9-3
Entrez Gene ID
75586
Gene Name
keratin associated protein 9-3
Gene Aliases
2310040M23Rik
Chromosome Location
Chr.11: 99597348 - 99598106 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029351.2 NP_083627.2 1 - 1 576 154
GenBank mRNA AK009730.1 - 1 - 1 578 154
AK131927.1 - 1 - 1 577 154

View Full Product Details