The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177753.3 | NP_808421.1 | 1 - 1 | 2860 | 150 |
GenBank mRNA | AY069926.1 | - | 1 - 1 | 1679 | 150 |
AY142959.1 | - | 1 - 1 | 1732 | 150 | |
BC063054.1 | - | 2 - 2 | 1648 | 150 | |
BC069176.1 | - | 2 - 2 | 2161 | 150 |