Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Oog1
- Entrez Gene ID
-
193322
- Gene Name
- oogenesin 1
- Gene Aliases
-
Oog, c-1
- Chromosome Location
-
Chr.12: 87552648 - 87608845 on Build GRCm38
- Gene Symbol
- Gm10436
- Entrez Gene ID
-
100039315
- Gene Name
- predicted gene 10436
- Gene Aliases
-
ENSMUSG00000072907
- Chromosome Location
-
Chr.12: 88175589 - 88210945 on Build GRCm38
- Gene Symbol
- Gm2042
- Entrez Gene ID
-
100039087
- Gene Name
- predicted gene 2042
- Gene Aliases
-
-
- Chromosome Location
-
Chr.12: 87954480 - 87960682 on Build GRCm38
View Full Product Details