The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113560.1 | NP_001107032.1 | 7 - 7 | 825 | 154 |
NM_025374.3 | NP_079650.3 | 6 - 6 | 1865 | 154 | |
GenBank mRNA | AK002386.1 | - | 6 - 6 | 1793 | 154 |
AK003567.1 | - | 6 - 6 | 1814 | 154 | |
AK037686.1 | - | - | 677 | 154 | |
AK049703.1 | - | 6 - 6 | 1822 | 154 | |
AK196705.1 | - | 1 - 1 | 207 | 154 | |
AK202257.1 | - | 1 - 1 | 116 | 154 | |
BC024663.1 | - | 6 - 6 | 1848 | 154 | |
BP769280.1 | - | 1 - 1 | 154 |