The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183221.3 | NP_899044.3 | 17 - 17 | 15975 | 163 |
GenBank mRNA | AK031348.1 | - | 1 - 1 | 2541 | 163 |
BC079887.1 | - | 9 - 9 | 7070 | 163 | |
DQ286572.1 | - | 17 - 17 | 15976 | 163 |