Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
Hba-a1
Entrez Gene ID
15122
Gene Name
hemoglobin alpha, adult chain 1
Gene Aliases
Hba, Hba1, Hbat1
Chromosome Location
Chr.11: 32283672 - 32284486 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008218.2 NP_032244.2 2 - 2 252 131
GenBank mRNA AK003077.1 - 2 - 2 250 131
AK010923.1 - 2 - 2 250 131
AK010927.1 - 2 - 2 251 131
AK010933.1 - 2 - 2 250 131
AK010937.1 - 2 - 2 253 131
AK010987.1 - 2 - 2 250 131
AK011018.1 - 2 - 2 252 131
AK011023.1 - 2 - 2 249 131
AK011025.1 - 2 - 2 252 131
AK011028.1 - 2 - 2 249 131
AK011055.1 - 2 - 2 253 131
AK011056.1 - - 788 131
AK011063.1 - 2 - 2 257 131
AK011076.1 - 2 - 2 250 131
AK011116.1 - - 763 131
AK012329.1 - 2 - 2 250 131
AK012755.1 - 2 - 2 250 131
AK171893.1 - 2 - 2 250 131
BC043020.1 - 2 - 2 232 131
L75940.1 - 2 - 2 131

Gene Symbol
Hba-a2
Entrez Gene ID
110257
Gene Name
hemoglobin alpha, adult chain 2
Gene Aliases
-
Chromosome Location
Chr.11: 32296492 - 32297310 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083955.1 NP_001077424.1 - 249 131
GenBank mRNA AK002222.1 - - 252 131
AK005434.1 - - 250 131
AK010422.1 - - 250 131
AK010938.1 - - 251 131
AK011030.1 - - 250 131
AK011073.1 - - 250 131
AK011121.1 - - 250 131
AK012348.1 - - 250 131
AK019138.1 - - 251 131
AK028173.1 - - 250 131
AK076068.1 - - 250 131
CF740193.1 - - 236 131
M26895.1 - - 223 131

View Full Product Details