The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009861.3 | NP_033991.1 | 6 - 6 | 1017 | 146 |
GenBank mRNA | AK051543.1 | - | 6 - 6 | 938 | 146 |
AK075567.1 | - | 6 - 6 | 936 | 146 | |
AK144216.1 | - | 7 - 7 | 942 | 148 | |
AK145659.1 | - | 5 - 5 | 781 | 146 | |
AK151087.1 | - | 6 - 6 | 962 | 146 | |
AK151726.1 | - | 6 - 6 | 939 | 146 | |
AK153564.1 | - | 6 - 6 | 961 | 146 | |
AK154870.1 | - | 6 - 6 | 937 | 146 | |
AK159470.1 | - | 6 - 6 | 979 | 146 | |
AK166281.1 | - | 6 - 6 | 936 | 148 | |
AK167195.1 | - | 6 - 6 | 925 | 146 | |
AK167400.1 | - | 6 - 6 | 930 | 146 | |
AK167609.1 | - | 6 - 6 | 935 | 146 | |
AK168013.1 | - | 6 - 6 | 931 | 147 | |
AK168076.1 | - | 6 - 6 | 929 | 146 | |
AK168089.1 | - | 6 - 6 | 931 | 147 | |
AK168503.1 | - | 6 - 6 | 947 | 146 | |
AK168758.1 | - | 6 - 6 | 970 | 146 | |
AK168820.1 | - | 6 - 6 | 943 | 146 | |
AK169122.1 | - | 6 - 6 | 951 | 146 | |
AK169232.1 | - | 6 - 6 | 930 | 146 | |
BC064792.1 | - | 6 - 6 | 923 | 146 | |
U37720.1 | - | 7 - 7 | 925 | 146 |