Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cbx1
Entrez Gene ID
12412
Gene Name
chromobox 1
Gene Aliases
Cbx, Cbx-rs2, E430007M08Rik, HP1B, Hp1beta, M31, MOD1
Chromosome Location
Chr.11: 96788679 - 96808982 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007622.3 NP_031648.1 6 - 6 973 128
GenBank mRNA AK014215.1 - 6 - 6 949 128
AK017608.1 - 5 - 5 713 128

View Full Product Details