The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011200.2 | NP_035330.1 | 1 - 1 | 54 | 167 |
GenBank mRNA | AK148288.1 | - | 1 - 1 | 55 | 167 |
CA315780.1 | - | 1 - 1 | 46 | 167 | |
U84411.1 | - | 1 - 1 | 44 | 167 |