The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134169.1 | NP_598930.1 | 1 - 1 | 564 | 130 |
GenBank mRNA | BC138428.1 | - | 1 - 1 | 613 | 130 |
BC138429.1 | - | 1 - 1 | 613 | 130 |