The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146293.1 | NP_001139765.1 | 11 - 12 | 1683 | 57 |
NM_133195.3 | NP_573458.2 | 11 - 12 | 1680 | 57 | |
GenBank mRNA | AK048427.1 | - | 11 - 12 | 1857 | 57 |
BC052744.1 | - | 11 - 12 | 1683 | 57 |