The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077361.1 | NP_001070829.1 | 7 - 8 | 1182 | 83 |
GenBank mRNA | AK158966.1 | - | 7 - 8 | 1110 | 83 |
BC055725.1 | - | 2 - 3 | 254 | 83 | |
BC059009.1 | - | 4 - 5 | 1462 | 83 |