Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pias3
Entrez Gene ID
229615
Gene Name
protein inhibitor of activated STAT 3
Gene Aliases
Pias3l
Chromosome Location
Chr.3: 96696375 - 96706070 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001165949.1 NP_001159421.1 8 - 9 1078 54
NM_018812.2 NP_061282.2 9 - 10 973 54
NM_146135.2 NP_666247.1 8 - 9 1115 54
GenBank mRNA AF034080.3 - 8 - 9 850 54
AK040619.1 - 8 - 9 1106 54
AK077551.1 - 8 - 9 1064 54
AK085503.1 - 8 - 9 1050 54
AK134404.1 - 3 - 4 253 54
BC023128.1 - 8 - 9 1021 54
BC050988.1 - 9 - 10 932 54
BC051252.1 - 8 - 9 988 54

View Full Product Details