Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ttc14
- Entrez Gene ID
-
67120
- Gene Name
- tetratricopeptide repeat domain 14
- Gene Aliases
-
2700016E08Rik, 4930434D01Rik, 4931403I22Rik, 4933402I15Rik, AI662468, AU014779, AW561908, cI-44, mKIAA1980
- Chromosome Location
-
Chr.3: 33800022 - 33814860 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK017763.1 |
- |
8 - 9 |
1475 |
77 |
View Full Product Details