The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175686.3 | NP_783617.2 | 4 - 5 | 1688 | 97 |
GenBank mRNA | AK008428.2 | - | 5 - 6 | 518 | 97 |
AK019503.1 | - | 4 - 5 | 499 | 97 | |
BC092372.1 | - | 4 - 5 | 1037 | 97 | |
L06502.1 | - | 4 - 5 | 853 | 97 | |
U03873.1 | - | 4 - 5 | 1694 | 97 |