The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172945.2 | NP_766533.2 | 13 - 14 | 1705 | 61 |
GenBank mRNA | AK081149.1 | - | - | 383 | 61 |
BC022693.1 | - | 3 - 4 | 417 | 61 | |
BC039786.1 | - | 3 - 4 | 890 | 61 | |
BC056928.1 | - | 8 - 9 | 934 | 61 | |
BC071189.1 | - | 7 - 8 | 791 | 61 | |
BC098207.1 | - | 6 - 7 | 744 | 61 | |
BC151158.1 | - | 13 - 14 | 1683 | 61 | |
BC172028.1 | - | 13 - 14 | 1684 | 61 |