The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083315.2 | NP_001076784.1 | 11 - 12 | 1479 | 87 |
NM_001289624.1 | NP_001276553.1 | 12 - 13 | 1472 | 87 | |
NM_001289625.1 | NP_001276554.1 | 12 - 13 | 1472 | 87 | |
NM_001289626.1 | NP_001276555.1 | 11 - 12 | 1403 | 87 | |
NM_001289627.1 | NP_001276556.1 | 12 - 13 | 1206 | 87 | |
NM_001289629.1 | NP_001276558.1 | 11 - 12 | 1137 | 87 | |
NM_022332.3 | NP_071727.2 | 12 - 13 | 1548 | 87 | |
GenBank mRNA | AF234885.1 | - | 11 - 12 | 1430 | 87 |
AK132103.1 | - | 11 - 12 | 1433 | 87 | |
AK151550.1 | - | 11 - 12 | 1434 | 87 | |
AY007792.1 | - | 12 - 13 | 1434 | 87 | |
AY007793.1 | - | 11 - 12 | 1365 | 87 | |
AY007794.1 | - | 13 - 14 | 1463 | 87 | |
AY007795.1 | - | 12 - 13 | 1394 | 87 | |
AY007796.1 | - | 13 - 14 | 1273 | 87 | |
AY007797.1 | - | 12 - 13 | 1204 | 87 | |
AY007798.1 | - | 12 - 13 | 1206 | 87 | |
AY007799.1 | - | 11 - 12 | 1137 | 87 | |
AY551821.1 | - | 12 - 13 | 1199 | 87 | |
AY551822.1 | - | 11 - 12 | 1130 | 87 | |
BC012719.1 | - | 6 - 7 | 619 | 87 | |
BC024652.1 | - | 12 - 13 | 1472 | 87 | |
BC051429.1 | - | 11 - 12 | 3158 | 87 | |
BC060630.1 | - | 12 - 13 | 3357 | 87 | |
BC108418.1 | - | 12 - 13 | 1492 | 87 | |
CT010245.1 | - | 11 - 12 | 1056 | 87 |