The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028421.1 | NP_082697.1 | 4 - 5 | 2124 | 91 |
NM_028864.2 | NP_083140.1 | 4 - 5 | 2124 | 91 | |
GenBank mRNA | AK004770.1 | - | 4 - 5 | 2125 | 91 |
AK020263.2 | - | 4 - 5 | 1803 | 91 | |
AK080334.1 | - | 4 - 5 | 2109 | 91 | |
AK143568.1 | - | 4 - 5 | 1794 | 91 | |
AK169402.1 | - | 4 - 5 | 1769 | 91 | |
AK172379.1 | - | 4 - 5 | 2125 | 91 |