Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Acsm4
Entrez Gene ID
233801
Gene Name
acyl-CoA synthetase medium-chain family member 4
Gene Aliases
O-MACS, OMACS
Chromosome Location
Chr.7: 119690026 - 119714566 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178414.3 NP_848501.1 11 - 12 1630 69
GenBank mRNA AK132127.1 - 11 - 12 1631 69
BC048390.1 - 11 - 12 1566 69

View Full Product Details