Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ikzf5
Entrez Gene ID
67143
Gene Name
IKAROS family zinc finger 5
Gene Aliases
2610034F18Rik, PEGASUS, Zfpn1a5
Chromosome Location
Chr.7: 131388649 - 131410506 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175115.4 NP_780324.2 1 - 2 94 92
GenBank mRNA AK033716.1 - 1 - 2 114 92
AK088289.1 - 1 - 2 137 92
AK169384.1 - 1 - 2 123 92
BC048183.1 - 1 - 2 94 92
BC058537.1 - 1 - 2 94 92
BC089455.1 - 1 - 2 116 93

View Full Product Details