Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clec5a
Entrez Gene ID
23845
Gene Name
C-type lectin domain family 5, member a
Gene Aliases
Clecsf5, Ly100, MDL-1, Mdl1
Chromosome Location
Chr.6: 40574898 - 40585805 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038604.1 NP_001033693.1 5 - 6 525 94
NM_021364.2 NP_067339.1 4 - 5 450 94
GenBank mRNA AF139769.1 - 4 - 5 420 94
AK036697.1 - 4 - 5 416 94
AK137482.1 - 4 - 5 451 94
BC104364.1 - 5 - 6 484 94
BC104365.1 - 5 - 6 484 94

View Full Product Details