Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Abhd16a
Entrez Gene ID
193742
Gene Name
abhydrolase domain containing 16A
Gene Aliases
AI326074, Bat-5, Bat5, D17H6S82E, NG26
Chromosome Location
Chr.17: 35089233 - 35102987 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178592.3 NP_848707.1 16 - 17 1407 61

View Full Product Details