Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pmel
Entrez Gene ID
20431
Gene Name
premelanosome protein
Gene Aliases
D10H12S53E, D12S53Eh, Pmel17, Si, Silv, gp100, gp87
Chromosome Location
Chr.10: 128705168 - 128720238 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021882.4 NP_068682.2 4 - 5 493 89
GenBank mRNA AK012808.1 - 4 - 5 494 89
AK131956.1 - 4 - 5 495 89
AK204124.1 - 2 - 3 176 89
BC075667.1 - 4 - 5 480 89
BC082555.1 - 4 - 5 479 89

View Full Product Details