Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Abcb8
Entrez Gene ID
74610
Gene Name
ATP-binding cassette, sub-family B (MDR/TAP), member 8
Gene Aliases
4833412N02Rik, AA409895
Chromosome Location
Chr.5: 24393681 - 24409947 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029020.2 NP_083296.2 3 - 4 903 62
GenBank mRNA AK014319.1 - 3 - 4 634 62
AK014688.1 - 3 - 4 656 62
AK030624.1 - 3 - 4 630 62
AK038760.1 - 3 - 4 630 62
AK049152.1 - 3 - 4 647 62
AK076315.1 - 3 - 4 654 62
AK166704.1 - 3 - 4 637 62
AK177107.1 - 1 - 2 145 62
AK177347.1 - 2 - 3 242 62
BC015301.1 - 3 - 4 903 62

View Full Product Details