The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028431.2 | NP_082707.1 | 12 - 13 | 1422 | 94 |
GenBank mRNA | AK013995.1 | - | 12 - 13 | 1423 | 94 |
AK152673.1 | - | 12 - 13 | 1424 | 94 | |
AK169417.1 | - | 12 - 13 | 1379 | 94 | |
BC038321.1 | - | 11 - 12 | 1293 | 94 | |
BC051216.1 | - | 11 - 12 | 1229 | 94 | |
BC060662.1 | - | 11 - 12 | 3934 | 94 | |
BC065095.1 | - | 11 - 12 | 3934 | 94 | |
BC087943.1 | - | 12 - 13 | 1434 | 94 |