The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164058.1 | NP_001157530.1 | 2 - 3 | 251 | 135 |
NM_008930.2 | NP_032956.1 | 2 - 3 | 248 | 135 | |
GenBank mRNA | AF011381.1 | - | 2 - 3 | 240 | 135 |
AF020525.1 | - | 2 - 3 | 222 | 135 | |
AK028371.1 | - | 2 - 3 | 249 | 135 | |
AK141309.1 | - | 2 - 3 | 252 | 135 | |
AK146050.1 | - | 2 - 3 | 252 | 135 | |
BB611867.1 | - | 2 - 3 | 249 | 135 | |
BC051671.1 | - | 2 - 3 | 233 | 135 |