The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130188.1 | NP_001123660.1 | 7 - 8 | 1053 | 66 |
NM_001130189.1 | NP_001123661.1 | 6 - 7 | 945 | 66 | |
NM_001130190.1 | NP_001123662.1 | 6 - 7 | 945 | 66 | |
NM_001130191.1 | NP_001123663.1 | 6 - 7 | 945 | 66 | |
NM_011360.3 | NP_035490.3 | 6 - 7 | 945 | 66 | |
GenBank mRNA | AB117975.1 | - | 6 - 7 | 778 | 66 |
AB200281.1 | - | 6 - 7 | 760 | 66 | |
AB200282.1 | - | 6 - 7 | 760 | 66 | |
AF031919.1 | - | 6 - 7 | 801 | 66 | |
AF103877.1 | - | 7 - 8 | 859 | 66 | |
AK076102.1 | - | 6 - 7 | 767 | 66 | |
AK132210.1 | - | 6 - 7 | 822 | 66 | |
BC012665.1 | - | 6 - 7 | 851 | 66 |