The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252266.1 | NP_001239195.1 | 1 - 2 | 95 | 86 |
NM_010492.3 | NP_034622.3 | 2 - 3 | 508 | 86 | |
GenBank mRNA | AK005965.1 | - | 2 - 3 | 213 | 86 |
AK016989.1 | - | 2 - 3 | 211 | 86 | |
BC002030.1 | - | 2 - 3 | 142 | 86 | |
BQ442697.1 | - | 2 - 3 | 508 | 86 | |
CF914747.1 | - | 2 - 3 | 211 | 86 | |
U26459.1 | - | 2 - 3 | 131 | 86 |